missing translation for 'onlineSavingsMsg'
Learn More

(2-Metilfenoxi)acetato de etilo, 99 %, Thermo Scientific™

Catalog Number p-7026399
Click to view available options
Cantidad:
5 g
25 g
100 g
Este artículo no se puede devolver. Vea la política de devoluciones
93917-68-1
C11H14O3
194.23
MFCD00216559
NHFQSFIBSJPLIW-UHFFFAOYSA-N
ethyl 2-methylphenoxy acetate, ethyl 2-2-methylphenoxy acetate, ethyl 2-o-tolyloxy acetate, ethyl 2-methylphenoxyacetate, ethyl-2-methylphenoxy acetate, o-tolyloxyacetic acid ethyl ester, o-tolyloxy-acetic acid ethyl ester, ethyl 2-2-methylphenoxy ethanoate
3022879
2-(2-metilfenoxi)acetato de etilo
CCOC(=O)COC1=CC=CC=C1C
Este artículo no se puede devolver. Vea la política de devoluciones
93917-68-1
C11H14O3
194.23
MFCD00216559
NHFQSFIBSJPLIW-UHFFFAOYSA-N
ethyl 2-methylphenoxy acetate, ethyl 2-2-methylphenoxy acetate, ethyl 2-o-tolyloxy acetate, ethyl 2-methylphenoxyacetate, ethyl-2-methylphenoxy acetate, o-tolyloxyacetic acid ethyl ester, o-tolyloxy-acetic acid ethyl ester, ethyl 2-2-methylphenoxy ethanoate
3022879
2-(2-metilfenoxi)acetato de etilo
CCOC(=O)COC1=CC=CC=C1C

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material Ethyl (2-methylphenoxy)acetate
Densidad 1.073
Punto de ebullición 119°C to 120°C
Temperatura de inflamación >110°C (230°F)
Índice de refracción 1.503
Cantidad 5 g
Formula Weight (peso de la fórmula) 194.23
Porcentaje de pureza 99%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'