missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-methyl-4-pentenoate, 98%

Product Code. 11424097
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11424097 5 g 5g
11434097 25 g 25g
11444097 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11424097 Supplier Thermo Scientific Alfa Aesar Supplier No. A19526.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 53399-81-8 | C8H14O2 | 142.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53399-81-8
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.20
MDL Number MFCD00010235
InChI Key BDBGKYIBDXAVMX-UHFFFAOYNA-N
Synonym ethyl 2-methyl-4-pentenoate, 2-methyl-4-pentenoic acid ethyl ester, ethyl 2-methyl 4-pentenoate, 4-pentenoic acid, 2-methyl-, ethyl ester, ethyl 2-methylpent-4-en-1-oate, ethyl 2-methyl-4-pentanoate, fema no. 3489, pineapple pentenoate, acmc-1ao14, ethyl-2-methyl-4-pentenoate
PubChem CID 62024
IUPAC Name ethyl 2-methylpent-4-enoate
SMILES CCOC(=O)C(C)CC=C

Specifications

Density 0.88
Boiling Point 153°C to 155°C
Flash Point 40°C (104°F)
Refractive Index 1.416
Quantity 5 g
UN Number UN3272
Beilstein 1753406
Formula Weight 142.2
Percent Purity 98%
Chemical Name or Material Ethyl 2-methyl-4-pentenoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.