missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-isocyanato-3-methylbutyrate, 97%, Thermo Scientific™

Product Code. 11342387
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
Product Code. Quantity unitSize
11342387 10 g 10g
11332387 2 g 2g
2 options
This item is not returnable. View return policy

Product Code. 11342387

Brand: Thermo Scientific Alfa Aesar L09988.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5296-78-6
Molecular Formula C8H13NO3
Molecular Weight (g/mol) 171.20
MDL Number MFCD00078478
InChI Key LXPOIQVGVGYFJA-UHFFFAOYNA-N
Synonym ethyl 2-isocyanato-3-methylbutyrate,97, ethyl 2s-2-isocyanato-3-methylbutanoate
PubChem CID 7018875
SMILES CCOC(=O)C(N=C=O)C(C)C

Specifications

Boiling Point 81°C to 82°C (3 mmHg)
Odor Irritating
Refractive Index 1.4275
Quantity 10 g
UN Number UN2206
Beilstein 1239965
Sensitivity Moisture sensitive
IUPAC Name ethyl 2-isocyanato-3-methylbutanoate
Formula Weight 171.2
Percent Purity 97%
Chemical Name or Material Ethyl 2-isocyanato-3-methylbutyrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.