missing translation for 'onlineSavingsMsg'
Learn More

2-Aminonicotinato de etilo, 98 %, Thermo Scientific Chemicals

Catalog Number 15442389
Click to view available options
Cantidad:
5 g
25 g
100 g
Este artículo no se puede devolver. Vea la política de devoluciones
13362-26-0
C8H10N2O2
166.18
MFCD01569452
KIAGEDYOPMHRRB-UHFFFAOYSA-N
12433260
2-aminopiridina-3-carboxilato de etilo
CCOC(=O)C1=C(N=CC=C1)N
Este artículo no se puede devolver. Vea la política de devoluciones
13362-26-0
C8H10N2O2
166.18
MFCD01569452
KIAGEDYOPMHRRB-UHFFFAOYSA-N
12433260
2-aminopiridina-3-carboxilato de etilo
CCOC(=O)C1=C(N=CC=C1)N

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material Ethyl 2-aminonicotinate
Punto de fusión 90°C to 96°C (literature)
Cantidad 25 g
Beilstein 124680
Sinónimo ethyl 2-aminonicotinate, 2-amino-nicotinic acid ethyl ester, ethyl2-aminonicotinate, 2-aminonicotinic acid ethyl ester, 3-pyridinecarboxylic acid, 2-amino-, ethyl ester, 2-amino nicotinic acid ethyl ester, 2-aminopyridine-3-carboxylic acid ethyl ester, pubchem12921, rarechem al bi 1234, buttpark 23\07-65
Formula Weight (peso de la fórmula) 166.18
Porcentaje de pureza 98%
Forma física Sólido

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'