Learn More
2-(2-amino-1,3-tiazol-4-il)acetato de etilo, Thermo Scientific™
Catalog Number p-4391334
Click to view available options
Cantidad:
25 g
50 g
100 g
53266-94-7
C7H10N2O2S
186.23
MFCD00005330
SHQNGLYXRFCPGZ-UHFFFAOYSA-N
ethyl 2-amino-4-thiazoleacetate, ethyl 2-2-aminothiazol-4-yl acetate, ethyl 2-2-amino-1,3-thiazol-4-yl acetate, ethyl 2-aminothiazole-4-acetate, ethyl 2-amino-1,3-thiazol-4-yl acetate, ethyl 2-aminothiazol-4-acetate, 2-amino-4-thiazolyl acetic acid ethyl ester, 4-thiazoleacetic acid, 2-amino-, ethyl ester, ethyl 2-amino-4-thiazolyl acetate
104454
2-(2-amino-1,3-tiazol-4-il)acetato de etilo
CCOC(=O)CC1=CSC(N)=N1
53266-94-7
C7H10N2O2S
186.23
MFCD00005330
SHQNGLYXRFCPGZ-UHFFFAOYSA-N
ethyl 2-amino-4-thiazoleacetate, ethyl 2-2-aminothiazol-4-yl acetate, ethyl 2-2-amino-1,3-thiazol-4-yl acetate, ethyl 2-aminothiazole-4-acetate, ethyl 2-amino-1,3-thiazol-4-yl acetate, ethyl 2-aminothiazol-4-acetate, 2-amino-4-thiazolyl acetic acid ethyl ester, 4-thiazoleacetic acid, 2-amino-, ethyl ester, ethyl 2-amino-4-thiazolyl acetate
104454
2-(2-amino-1,3-tiazol-4-il)acetato de etilo
CCOC(=O)CC1=CSC(N)=N1
Nombre del producto químico o material Ethyl 2-(2-amino-1, 3-thiazol-4-yl)acetate
Intervalo de porcentaje de ensayo 97%
Envase Frasco de vidrio ámbar
Cantidad 25 g
Formula Weight (peso de la fórmula) 186.23
Porcentaje de pureza 97%