missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Boc-3-oxopiperidina-4-carboxilato de etilo, 97 %, Thermo Scientific Chemicals
Click to view available options
Cantidad:
250 mg
1 g
5 g
Descripción
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Especificaciones
Nombre del producto químico o material | Ethyl 1-Boc-3-oxopiperidine-4-carboxylate |
Cantidad | 250 mg |
Beilstein | 421506 |
Formula Weight (peso de la fórmula) | 271.31 |
Porcentaje de pureza | 97% |
Forma física | Líquido |
RUO - Para uso exclusivo en investigación
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'