Learn More
Ether, Preserved with about 2% Alcohol, 0.2% max Water and 7ppm min BHT, U.S.P., J.T. Baker™
Catalog Number 15558554
Click to view available options
Cantidad:
250 mL
1 L
Envase:
Frasco de aluminio
60-29-7
C4H10O
74.12
MFCD00011646
RTZKZFJDLAIYFH-UHFFFAOYSA-N
diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
3283
CHEBI:35702
etoxietano
CCOCC
60-29-7
C4H10O
74.12
MFCD00011646
RTZKZFJDLAIYFH-UHFFFAOYSA-N
diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
3283
CHEBI:35702
etoxietano
CCOCC
Nombre del producto químico o material Ether
Nombre de nota Preserved with about 2% Alcohol, 0.2% max Water and 7ppm min BHT
Temperatura de inflamación −45.00°C
Envase Frasco de aluminio
Cantidad 250 mL
Gravedad específica 1L = 0.71kg
Formula Weight (peso de la fórmula) 74.12
Grado USP