missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERCC3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 10580-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
The human XPB DNA helicase is a subunit of the DNA repair/basal transcription factor TFIIH, and is involved in early steps of the nucleotide excision repair (NER) pathway. Two distinct clinical phenotypes, xeroderma pigmentosum associated with Cockayne′s syndrome (XP/CS) and trichothiodystrophy (TTD), can be due to mutations in the XPB gene.Especificaciones
ERCC3 | |
Polyclonal | |
Unconjugated | |
ERCC3 | |
BTF2, BTF2 p89, ERCC3, GTF2H, RAD25, TFIIH, TFIIH 89 kDa subunit, TFIIH p89, XPB, XPBC | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
13872, 2071, 291703 | |
-20°C | |
Liquid |
Western Blot, Immunoprecipitation | |
0.26 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P19447, P49135, Q4G005 | |
Ercc3 | |
ERCC3 Fusion Protein Ag0876 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |