Learn More
ER beta/NR3A2 Antibody (6B12), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 5 publications
Marca:  Novus Biologicals NB200-304
Detalles adicionales : Peso : 0.00970kg
Código de producto. 18215430
  • 535.62€ / 100 microlitros
Fecha estimada de envío: 14-08-2024
para ver el stock
Descripción
Descripción
ER beta/NR3A2 Monoclonal specifically detects ER beta/NR3A2 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Especificaciones
Especificaciones
ER beta/NR3A2
Monoclonal
1mg/mL
Western Blot 1:500-1:3000, Immunohistochemistry 1:10 - 1:500
Erb, ER-beta, ESR-BETA, ESTRB, estrogen receptor 2 (ER beta), estrogen receptor beta, estrogen receptor beta 4, NR3A2ESRB, Nuclear receptor subfamily 3 group A member 2
Mouse
59 kDa
100 μL
Cancer, GPCR
2100
Human
Purified
Western Blot, Immunohistochemistry
6B12
Unconjugated
Q92731
ESR2
Amino acids 1-153 of human ER expressed in E. coli.
Protein G purified
RUO
Primary
Estrogen Receptor beta (6B12). NB 200-304 reacts with human ER-b.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG1