Learn More
Eppendorf™ ep Dualfilter T.I.P.S.™ Puntas de pipeta con filtro SealMax
Las puntas con tecnología de bloqueo de líquidos ofrecen una protección adicional durante el pipeteo
Marca: Eppendorf™ 0030077857
Detalles adicionales : Peso : 2.02288kg
Descripción
- Sterile tips made of polyethylene
- White and violet filter layers in the tip retain aerosols and biomolecules
- Purple filter layer blocks liquids, binds aerosols and does not allow any liquid to pass through
- Hydrophobic white layer repels drops, splashes and binds aerosols
- Free of PCR inhibiting additives
- Provide protection against contamination of pipet and sample
- Color coded tip boxes simplify matching pipetes with their tips
PCR and microbiology procedures
Especificaciones
Rack | |
Filtered | |
Azul | |
7.6 cm |
De 50 a 1000 μl | |
Sí | |
Polietileno |