Learn More
EPHA1 Recombinant Human Protein
EPHA1 Recombinant Human Protein
Marca:  Invitrogen PV3841
Detalles adicionales : Peso : 0.05250kg
Código de producto. 11533577
  • 634.00€ / 10 microgramos
Fecha estimada de envío: 16-04-2024
para ver el stock
Descripción
Descripción
EPHA1 belongs to the ephrin receptor subfamily of the protein tyrosine kinase family. EPH and EPH-related receptors have been implicated in mediating developmental events, particularly in the nervous system.
Especificaciones
Especificaciones
NP_005223.2
Pharma and Biopharma, Protein Assays and Analysis, Target and Lead Identification and Validation, Kinase Assay
74.4kDa
10μg
For research use only. Not for use in diagnostic procedures
EPHA1, EPH
EPHA1 was pre-diluted in enzyme dilution buffer and assayed in 25mM Tris (pH 7.5), 10mM MgCl2, 0.5mM EGTA, 0.5mM Na3VO4, 5mM β-glycerophosphate, 2.5mM DTT, 0.01% Triton™ X-100, 200μM ATP, 200μg/mL poly [Glu, Tyr] 4:1 substrate and trace [32P]-γ-ATP for 10 minutes at 30°C
Amino acids 568-976
220nmoles of phosphate transferred to poly [Glu, Tyr] 4:1 substrate per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 2μg/mL
GST Tag
Kinase and inhibitors
90% by SDS-PAGE
0.35mg/mL
2041
EPHA1 Recombinant Human Protein
–80°C
1 Each
EPHA1 Recombinant Human Protein
Ephrin and EPH receptor
EPHA1 Recombinant Human Protein
Human
Baculovirus
Receptor tyrosine kinases
Sugerencias de productos
Sugerencias de productos
Videos
SDS
Documentos
Documentos
Certificados
Ofertas especiales
Ofertas especiales
For Research Use Only. Not for use in diagnostic procedures.