Learn More
EPHA1 Recombinant Human Protein
EPHA1 Recombinant Human Protein
Marca:  Invitrogen PV3841
Detalles adicionales : Peso : 0.05250kg
 Ver más versiones de este producto
Código de producto. 11533577
  • 464.00€ / 10 microgramos
Fecha de envío estimada
del proveedor 04-06-2019
Añadir a la cesta
Descripción
Descripción
EPHA1 belongs to the ephrin receptor subfamily of the protein tyrosine kinase family. EPH and EPH-related receptors have been implicated in mediating developmental events, particularly in the nervous system.
Especificaciones
Especificaciones
NP_005223.2
Pharma and biopharma, protein assays and analysis, target and lead identification and validation
74.4kDa
90% by SDS-PAGE
–80°C
1 Each
EPHA1, EPH
EPHA1 was pre-diluted in enzyme dilution buffer and assayed in 25mM Tris (pH 7.5), 10mM MgCl2, 0.5mM EGTA, 0.5mM Na3VO4, 5mM β-glycerophosphate, 2.5mM DTT, 0.01% Triton™ X-100, 200μM ATP, 200μg/mL poly [Glu, Tyr] 4:1 substrate and trace [32P]-γ-ATP for 10 minutes at 30°C
Kinase and inhibitors
Ephrin and EPH receptor
EPHA1 Recombinant Human Protein
Human
0.35mg/mL
2041
EPHA1 Recombinant Human Protein
10μg
For research use only. Not for use in diagnostic procedures
Receptor tyrosine kinases
EPHA1 Recombinant Human Protein
Kinase assay
Baculovirus
Amino acids 568-976
220nmoles of phosphate transferred to poly [Glu, Tyr] 4:1 substrate per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 2μg/mL
GST Tag
Videos
For Research Use Only. Not for use in diagnostic procedures.