missing translation for 'onlineSavingsMsg'
Learn More

EPAS1 Mouse anti-Human, Clone: 2F6B12, Proteintech

Mouse Monoclonal Antibody

Marca:  Proteintech 66731-1-IG-150UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16818885

  • 319.00€ / 150 microlitros

Envío estimado: 02-11-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

HIF-2 alpha (Epas1) is a transcription factor involved in the induction of genes regulated by oxygen. HIF-2 alpha contains a basic-helix-loop-helix domain protein dimerization domain as well as a domain found in proteins in signal transduction pathways which respond to oxygen levels. It also regulates the vascular endothelial growth factor (VEGF) expression and seems to be implicated in the development of blood vessels and the tubular system of the lungs. Mutations in this gene are associated with erythrocytosis familial type 4.
Especificaciones

Especificaciones

EPAS1
2F6B12
Unconjugated
EPAS1
Liquid
EPAS1
EPAS1 Fusion Protein Ag24886
Protein A
RUO
Antibody
Monoclonal
Human
Western Blot
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q99814
bHLHe73, ECYT4, EPAS 1, EPAS1, HIF 1 alpha like factor, HIF 2 alpha, HIF2 alpha, HIF2A, HLF, Member of PAS protein 2, MOP2, PASD2
Mouse
IgG2a
150 μL
-20°C
Primary
2034
Videos
SDS
Documentos
Certificados