Learn More
Entecavir Monohydrate, TRC
Product Code. 30385259
Change view
Click to view available options
Quantity:
2 mg
Unit Size:
2mg
Product Code. Quantity Unit Size
30385259 2 mg 2mg
1 options
This item is not returnable. View return policy
Product Code. 30385259 Supplier Toronto Research Chemicals Supplier No. TRCE5589482MG
 Restricted Item
Fecha estimada de envoi: 26-04-2027
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Entecavir Monohydrate
CAS 209216-23-9
Molecular Formula C12 H15 N5 O3 . H2 O
Quantity 2 mg
Synonym 6H-Purin-6-one, 2-amino-1,9-dihydro-9-[(1S,3R,4S)-4-hydroxy-3-(hydroxymethyl)-2-methylenecyclopentyl]-, hydrate (1:1), 6H-Purin-6-one, 2-amino-1,9-dihydro-9-[(1S,3R,4S)-4-hydroxy-3-(hydroxymethyl)-2-methylenecyclopentyl]-, monohydrate (9CI), Entecavir monohydrate
InChI Formula InChI=1S/C12H15N5O3.H2O/c1-5-6(3-18)8(19)2-7(5)17-4-14-9-10(17)15-12(13)16-11(9)20;/h4,6-8,18-19H,1-3H2,(H3,13,15,16,20);1H2/t6-,7-,8-;/m0./s1
SMILES O.NC1=Nc2c(ncn2[C@H]3C[C@H](O)[C@@H](CO)C3=C)C(=O)N1
IUPAC Name 2-amino-9-[(1S,3R,4S)-4-hydroxy-3-(hydroxymethyl)-2-methylidenecyclopentyl]-1H-purin-6-one;hydrate
Molecular Weight (g/mol) 295.29
Formula Weight 295.1281