missing translation for 'onlineSavingsMsg'
Learn More

ENPP-1 Antibody, Biotin, Novus Biologicals™

Goat Polyclonal Antibody

Marca:  Bio-Techne NB600-816B

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18214193

  • 519.42€ / 0.10 ml
Fecha estimada de envío: 14-08-2024
para ver el stock

Descripción

Descripción

ENPP-1 Polyclonal specifically detects ENPP-1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.
Especificaciones

Especificaciones

ENPP-1
Polyclonal
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA
P22413
ENPP1
A synthetic peptide made to a C-terminal portion of the human ENPP1 protein (between residues 900-925) [UniProt P22413]
Affinity Purified
RUO
Primary
Human. Immunogen sequence has 85% homology to mouse and rat.
Store at 4C in the dark.
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA
Biotin
PBS with 0.05% Sodium Azide
ectonucleotide pyrophosphatase/phosphodiesterase 1, E-NPP 1, Ly-41 antigen, M6S1NPP1, Membrane component chromosome 6 surface marker 1, membrane component, chromosome 6, surface marker 1, NPPSalkaline phosphodiesterase 1, PC1, PC-1, PCA1ARHR2, PDNP1ectonucleotide pyrophosphatase/phosphodiesterase family member 1, Phosphodiesterase I/nucleotide pyrophosphatase 1, plasma-cell membrane glycoprotein 1, Plasma-cell membrane glycoprotein PC-1
Goat
140 kDa
0.1 mL
Lipid and Metabolism
5167
Human
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales

For Research Use Only