Learn More
Enoxaparin Sodium, TRC
Product Code. 30138088
Change view
Click to view available options
Quantity:
25 mg
50 mg
200 mg
Unit Size:
200mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138088 50 mg 50mg
30138379 25 mg 25mg
30004397 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138088 Supplier Toronto Research Chemicals Supplier No. TRCH24580850MG
 Restricted Item
Fecha estimada de envoi: 17-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 679809-58-6
Molecular Formula C42H59N3Na4O35S2
Molecular Weight (g/mol) 1322
Synonym Clexane, Enoxaparin, Enoxil, Inhixa, Jipailin, Klexane, Lovenox, PK 10169, Thorinane, Xaparin;
IUPAC Name sodium (2R,3R,4S)-2-(((2R,3S,4R,5R,6S)-5-acetamido-6-(((1R,2R,3R,4R)-4-(((2R,3S,4R,5R,6R)-5-acetamido-6-(((4R,5R,6R)-2-carboxylato-4,5-dihydroxy-6-(((1R,3R,4R,5R)-3-hydroxy-4-(sulfonatoamino)-6,8-dioxabicyclo[3.2.1]octan-2-yl)oxy)tetrahydro-2H-pyran-3-yl)oxy)-2-(hydroxymethyl)-4-methyltetrahydro-2H-pyran-3-yl)oxy)-6-carboxylato-2,3-dihydroxycyclohexyl)oxy)-4-hydroxy-2-((sulfooxy)methyl)tetrahydro-2H-pyran-3-yl)oxy)-3,4-dihydroxy-3,4-dihydro-2H-pyran-6-carboxylate
SMILES C[C@@H]1[C@H]([C@H](O[C@@H]([C@H]1O[C@@H]2CC([C@H]([C@@H]([C@H]2O)O)O[C@@H]3[C@@H]([C@H]([C@@H]([C@H](O3)COS(=O)(=O)O)O[C@H]4[C@@H]([C@H](C=C(O4)C(=O)[O-])O)O)O)NC(=O)C)C(=O)[O-])CO)OC5[C@@H]([C@H]([C@@H](OC5C(=O)[O-])OC6[C@H]7CO[C@H](O7)[C@@H]([C@H]6O)NS(=O)(=O)[O-])O)O)NC(=O)C.[Na+].[Na+].[Na+].[Na+]
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C42H63N3O35S2.4Na/c1-9-19(43-10(2)47)39(79-33-27(55)28(56)42(80-34(33)37(61)62)78-31-17-7-69-38(74-17)21(25(31)53)45-81(63,64)65)73-16(6-46)29(9)71-14-4-12(35(57)58)30(26(54)23(14)51)76-40-20(44-11(3)48)24(52)32(18(75-40)8-70-82(66,67)68)77-41-22(50)13(49)5-15(72-41)36(59)60;;;;/h5,9,12-14,16-34,38-42,45-46,49-56H,4,6-8H2,1-3H3,(H,43,47)(H,44,48)(H,57,58)(H,59,60)(H,61,62)(H,63,64,65)(H,66,67,68);;;;/q;4*+1/p-4/t9-,12?,13+,14-,16-,17-,18-,19-,20-,21-,22-,23+,24-,25-,26-,27-,28-,29+,30-,31?,32-,33?,34?,38-,39-,40-,41+,42-;;;;/m1..../s1
Formula Weight 1321.2
Chemical Name or Material Enoxaparin Sodium