missing translation for 'onlineSavingsMsg'
Learn More

eNOS Antibody, Alexa Fluor™ 532, Novus Biologicals™

Rabbit Polyclonal Antibody

Marca:  Bio-Techne NB300-500AF532

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18213775

  • 536.70€ / 0.10 ml
Fecha estimada de envío: 21-08-2024
para ver el stock

Descripción

Descripción

eNOS Polyclonal specifically detects eNOS in Human, Mouse, Rat, Porcine, Amphibian, Bovine, Canine, Drosophila, Sheep samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Especificaciones

Especificaciones

eNOS
Polyclonal
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Constitutive NOS, EC 1.14.13.39, ECNOS, endothelial, Endothelial NOS, eNOScNOS, nitric oxide synthase 3 (endothelial cell), NOS type III, NOSIII
Rabbit
0.1 mL
4846
Human, Mouse, Rat, Porcine, Amphibian, Bovine, Canine, Drosophila, Sheep
IgG
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Alexa Fluor 532
50 mM sodium borate with 0.05% sodium azide
NOS3
Synthetic peptide corresponding to residues S(1179) LQERQLRGAVPWAFD(1194) of human eNOS.
Primary
This does not detect human inducible NOS or rat brain NOS.
Store at 4°C in the dark.
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales