missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ ENO2 (Human) ELISA Kit

Enzyme Immunoassay for the quantitative measurement of ENO2 in Serum

Marca:  Abnova™ KA2122.1Kit

Detalles adicionales : Peso : 0.02000kg

 Ver más versiones de este producto

Código de producto. 16057045

  • 578.00€ / 1 set


Fecha estimada de envío
18-02-2020
Añadir a la cesta

Especificaciones

Especificaciones

1.25 to 80ng/mL
Human
0.15ng/mL
Quantification
ENO2 (Human) ELISA Kit
1 Kit
Buffered Solution, Cell Lysate, Serum, Plasma
96 Assays
HRP
ENO2 (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human ENO2.
Colorimetric
96 Wells antibody coated microwells, Incubation Buffer (30mL), Washing Buffer (10X) (100mL), Standard Protein (lyophilized) (1 Glass vial), Standard/Sample/ Secondary Antibody Dilution Buffer (25mL), Secondary Antibody (lyophilized) (1 Glass vial), AV-HRP (150μL), AV-HRP Dilution Buffer (25mL), TMB Substrate (20mL), Stop Solution (20mL)
ENO2 (Human) ELISA kit
For research use only (RUO)
Store the kit at 4°C.
100μL
Videos
Documentos