Learn More
Merck™ Vent Filters S.D.S Systems (purchased after 12/30/03)
Protect high-resistivity water product from ingress of possible airborne contaminants from the environment into S.D.S tank during storage
Marca:  Merck™ TANKVNT01
Detalles adicionales : Peso : 2.00000kg
 Ver más versiones de este producto
Código de producto. 15270159
  • 474.00€ / cada uno
Fecha de envío estimada
del proveedor 04-03-2020
Añadir a la cesta
Descripción
Descripción
  • Filter incorporates activated carbon to absorb volatile organics, soda lime bed to remove carbon dioxide, and an absolute membrane to remove particles and bacteria from the laboratory atmosphere
  • Filters easily install in built-in holder on front of S.D.S. reservoir
Note:These filters are for use with S.D.S. systems purchased after December 31, 2003; for S.D.S systems purchased before December 31, 2003, contact your Fisher Scientific™ Customer Service Representative
Especificaciones
Especificaciones
Vent Filter
Vent Filter
RiOs™ system