Learn More
Merck™ Filter Holder Vacuum Manifolds
Provide vacuum support for simultaneous filtration of either three or six test samples
Marca:  Merck™ XX2504700
Detalles adicionales : Peso : 0.20000kg
 Ver más versiones de este producto
Código de producto. 11522124
  • 4545.00€ / cada uno
Consúltenos : 902 239 303 
Añadir a la cesta
Descripción
Descripción
  • Use Millipore glass, plastic or stainless-steel filter holders fitted with standard No. 8 perforated stoppers over the holder outlets
  • Manifolds connect to single vacuum source and use separate control valves at each station for independent operation
  • Use vent ports on each stainless-steel manifold station to attach Swinnex™ filter holders or Millex™ filter units to maintain sterility on the test filter's downstream side
Stainless-steel Manifolds
  • Stainless-steel body, three-way valves, filter holder supports; anodized aluminum handles; stainless-steel hose connector, pipe plug
  • Units are autoclavable at 121°C (250°F)
PVC Manifold
  • Polyvinylchloride body; stainless-steel two-way valve; polypropylene hose adapters and handles
  • Not autoclavable
Especificación
Especificación
Manifold
Stainless Steel
Vacuum Filter Holder
Videos
Documentación
Documentos técnicos