Learn More
Merck Chemicon™ EndoGRO™-MV Complete Culture Media Kit
Product Code. 17826897 Shop All Merck Products
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
Product Code. Quantity Unit Size
17826897 500 mL 500mL
1 options
This item is not returnable. View return policy
Product Code. 17826897 Supplier Merck Supplier No. SCME004
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
Currently Unavailable: We have no information when this product will be available.
View alternative products
This item is not returnable. View return policy
a 5% serum formulation for culturing human microvascular endothelial cells, this media does not contain VEGF.
EndoGRO Media Formulations provide an optimal cell culture environment for many types of endothelial cells, including HUVEC, aortic endothelial cells and other human large vessel and microvascular endothelial cells. These media have been shown to grow endothelial cells at rates that meet or exceed commercially available serum containing media, while maintaining excellent cell morphology. The media are packaged in a specially designed UV protective shrinkwrap for added stability, which includes a temperature gauge that helps prevent contamination. EndoGRO Media contain no antimicrobials and no phenol red, components that can cause cell stress and masking effects that may influence experimental results. Four different media formulations are available for various applications and cell culture requirements.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Cell culture
Quantity 500 mL
Content And Storage Basal medium: 2-8°C in dark; Supplement Mix: -20