Learn More
Elma™ Elmasonic Noise Protection Covers
Product Code. 17861347
Change view
Click to view available options
Model:
500
900
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model Unit Size
17861347 900 Each
17841297 500 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17861347 Supplier Elma™ Supplier No. 1100292
  • ONLINE EXCLUSIVE PRICE
    710.00€
    536.97€ / paquete de 1
    Ahorro 173.03€
    24% Descuento
Fecha estimada de envoi: 30-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Elmasonic Noise Protection Covers save energy and costs by reducing waste heat and collecting and recirculating the water vapor.
  • Compatible covers in various designs are available for each ultrasonic cleaning unit
  • Cover performs a variety of functions
  • Reduces the noise level during the cleaning process by up to 54%
  • Underside of the cover serves as a catch tray for dripping water – simply place the Elma Basket on the upturned cover once the cleaning process is complete
  • Cover also returns evaporating water to the ultrasonic bath to save energy and reduce costs
  • Energy and cost savings
  • Can be used as a drainer
Specifications
Product Type Noise Protection Cover
For Use With (Equipment) Fisherbrand Select, Fisherbrand EASY, Elmasonic Med, Elmasonic Select
Model 900