Learn More
Electrode Filling Solution, 3M KCl, Reagecon™
BIOPHARMA
Product Code. 11543162 Shop All Reagecon Products
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Product Code. Quantity Unit Size
11543162 100 mL 100mL
1 options
This item is not returnable. View return policy
Product Code. 11543162 Supplier Reagecon Supplier No. EFS3
  • ONLINE EXCLUSIVE PRICE
    55.31€
    50.56€ / 100 ml
    Ahorro 4.75€
    8% Descuento
View all promotions
 Restricted Item
En stock
Add to basket
Edge
Add to basket
This item is not returnable. View return policy
Suitable for all Blueline electrodes. Electrode Filling Solution, 3M KCl, Reagecon™ is a ready-to-use potassium chloride solution without silver ions. It is used to store pH electrodes when they are not in use to help ensure that electrodes produce fast response times and accurate results.
Glass electrodes need to be kept moist. If an electrode dries out during storage, a regeneration procedure is required to restore the hydrated glass layer and the reference junction in order to make the electrode operable. Filling the storage cap/tube with a potassium chloride solution keeps the electrode hydrated to maximize electrode performance and extend electrode life.
Specifications
Color Colorless
Composition KCl in Water
Concentration 3 mol/L
For Use With (Equipment) All Blueline pH Electrodes
Type Electrode Filling Solution
Filling Solution Type 3M KCl without Silver Ions
Odor Odorless
Quantity 100 mL