Learn More
SI Analytics™ Electrode Double Clamp
Product Code. 10413891 Shop All SI Analytics Products
Change view
Click to view available options
For Use With (Equipment):
Stand Z 351, Holder for Two Electrodes or One Electrode and One Titration Tip
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
10413891 Stand Z 351, Holder for Two Electrodes or One Electrode and One Titration Tip Each
1 options
This item is not returnable. View return policy
Product Code. 10413891 Supplier SI Analytics™ Supplier No. 285123082
  • ONLINE EXCLUSIVE PRICE
    64.09€
    58.00€ / paquete de 1
    Ahorro 6.09€
    9% Descuento
View all promotions
Fecha estimada de envoi: 23-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
SI Analytics™ Electrode Double Clamp is designed to hold two electrodes or one electrode and one titration tip.
  • Holds two electrodes
  • Capable of holding one electrode and one titration tip
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Stand Z 351, Holder for Two Electrodes or One Electrode and One Titration Tip
Type Electrode Double Clamp