Learn More
Invitrogen™ EGFR (ErbB1) [T790M] Recombinant Human Protein
The epidermal growth factor receptor is a receptor tyrosine kinases that signals in response to various growth factors.
Marca: Invitrogen™ PR8052A
Detalles adicionales : Peso : 0.01000kg
Descripción
Overexpression of EGFR has been linked to numerous types of cancer and EGFR is the target of both biological and small molecular therapeutics. The T790M mutant of EGFR has arisen in the clinics and displays resistance to Erlotinib (Tarceva) and Gefitinib (Iressa).
Especificaciones
NP_005219.2; T790M | |
50mM Tris pH 7.5, 150mM NaCl, 0.02% Trition X-100, 0.5mM EDTA, 2mM DTT, 50% Glycerol | |
100μg | |
RUO - research use only | |
EGFR (ErbB1) T790M | |
Human | |
GST Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
90.5kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Cytoplasmic | |
EGF Receptor (ErbB) Kinase Family | |
Recombinant | |
Baculovirus |