missing translation for 'onlineSavingsMsg'
Learn More

EGF Antibody (RM0078-2M13), FITC, Novus Biologicals™

Rat Monoclonal Antibody

Marca:  Bio-Techne NBP1-21708F

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18215941

  • 590.70€ / 0.10 ml
Fecha estimada de envío: 19-08-2024
para ver el stock

Descripción

Descripción

EGF Monoclonal specifically detects EGF in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Especificaciones

Especificaciones

EGF
Monoclonal
FITC
PBS with 0.05% Sodium Azide
EGF
Mouse recombinant EGF
0.1 mL
Primary
This was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with mouse recombinant EGF.
Store at 4C in the dark.
IgG2
Immunohistochemistry, Immunohistochemistry (Paraffin)
RM0078-2M13
Immunohistochemistry, Immunohistochemistry-Paraffin
beta-urogastrone, epidermal growth factor, epidermal growth factor (beta-urogastrone), hEGF, HOMG4, pro-epidermal growth factor, URG
Rat
Protein A or G purified
Angiogenesis, Biologically Active Proteins, Cancer, Cell Cycle and Replication, Cytokine Research, Immunology, Virology Bacteria and Parasites
1950
Mouse
Purified
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales