Learn More
Cole-Parmer™ Economical Drive
Product Code. 15229159 Shop All Masterflex Products
Change view
Click to view available options
Speed:
150 to 4000 rpm
Unit Size:
Each
Product Code. Speed Unit Size
15229159 150 to 4000 rpm Each
1 options
This item is not returnable. View return policy
Product Code. 15229159 Supplier Cole-Parmer™ Supplier No. 0700390
  • ONLINE EXCLUSIVE PRICE
    2675.00€
    2025.68€ / paquete de 1
    Ahorro 649.32€
    24% Descuento
Fecha estimada de envoi: 05-02-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Order Info
TemperatureMetricRange
Specifications
Electrical Requirements 0 to 24 VDC
Max. Operating Temperature (Metric) 40°C
Motor Type PM brush, TENV
Min. Operating Temperature (Metric) 4°C
Overall Height (Metric) 7.6 cm
Overall Length (Metric) 10.1 cm
Overall Width (Metric) 7.6 cm
Speed 150 to 4000 rpm