Learn More
Velp Scientifica™ ECO 6 COD Test Tubes, Pk of 3
Product Code. 17129170 Shop All Velp Products
Change view
Click to view available options
Volume (Metric):
200 mL
Unit Size:
Pack of 3
Product Code. Volume (Metric) Unit Size
17129170 200 mL Pack of 3
1 options
This item is not returnable. View return policy
Product Code. 17129170 Supplier Velp Scientifica™ Supplier No. A00000145
  • ONLINE EXCLUSIVE PRICE
    94.60€
    77.50€ / paquete de 3
    Ahorro 17.10€
    18% Descuento
Fecha estimada de envoi: 23-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Velp Scientifica™ ECO 6 COD Test Tube for use with ECO 6 Thermoreactor.
  • For use with ECO 6 Thermoreactor
Specifications
Volume (Metric) 200 mL
Format Test Tube
Includes NS 29/32
Quantity 3 Tubes
Diameter (Metric) Outer 42 mm