missing translation for 'onlineSavingsMsg'
Learn More

ECD Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 10192-1-AP-150UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16816413

  • 319.00€ / 150 microlitros

Envío estimado: 21-10-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

ERp72 is an endoplasmic reticulum luminal protein that is both a stress protein and a member of the protein disulfide isomerase family of proteins. Sequence analysis of ERp72 shows that it shares sequence identity with PDI (protein disulphide isomerase) at three discrete regions, as well as containing the ER retention signal KEEL at its C-terminus. ERp72 is shown to have three copies of sequence believed to be CGHC-containing active sites, which are also found in PDI. These CGHC sites are critical sites that are involved in the protein disulphide isomerization process. These results suggest that ERp72 is a newly described member of the family of CGHC-containing proteins that functions as a molecular chaperone.
Especificaciones

Especificaciones

ECD
0.31 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O95905, Q9CS74
ECD, GCR2, HSGT1, Protein ecdysoneless homolog, Protein SGT1, SGT1, Suppressor of GCR2
Rabbit
IgG
150 μL
-20°C
Primary
11319, 70601
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Unconjugated
ECD
Liquid
Ecd
ECD Fusion Protein Ag0245
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Videos
SDS
Documentos
Certificados