Learn More
Fisherbrand™ Ebony Treatment Dish
Product Code. 15599901 Shop All Fisherbrand Electrochemistry Products
Change view
Click to view available options
No. per Pack:
1 Each
4
Unit Size:
Each
Pack of 4
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. per Pack Unit Size
15599901 4 Pack of 4
15589901 1 Each Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15599901
  • ONLINE EXCLUSIVE PRICE
    319.00€
    313.77€ / paquete de 4
    Ahorro 5.23€
    1% Descuento
Fecha estimada de envoi: 28-12-2026
Add to basket
Add to basket
Protects DNA from damage. Fisherbrand™ Ebony Treatment Dishes are manufactured with a darkened acrylic to reduce exposure to light that may be harmful to DNA.
  • Manufactured from Ebony Acrylic
  • Minimizes exposure to light and reduces background DNA damage
Specifications
Color Ebony
For Use With (Equipment) Comet Assay Tank
Material Acrylic
No. per Pack 4