Learn More
Hettich Lab Technology™ EBA 280 S Centrifuge
Product Code. 15321804
Change view
Click to view available options
Capacity:
6 x 50 mL
Product Code. Capacity
15321804 6 x 50 mL
1 options
This item is not returnable. View return policy
Product Code. 15321804 Supplier Hettich Lab Technology™ Supplier No. 1102
 Restricted Item
Fecha estimada de envoi: 14-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
EBA 280 S Centrifuge offers exceptional user comfort and a choice of 6 rotors. The rotor can be removed and replaced easily thanks to the quick release system. The rotor locks in place and remains secure without the use of tools or extra effort. The backlit digital control panel offers the opportunity to save 9 multiple program settings.
  • Plastic housing
  • Metal lid
  • Viewing port in the lid
  • One-hand lid lock
  • Lid dropping protection
  • Emergency lid lock release
  • Stainless steel chamber
  • Automatic rotor recognition
  • Brushless drive
  • Error display
  • Imbalance monitoring and switch-off
  • Backlit panel with actual values of all parameters
  • 3 adjustable messages after completion of the centrifugation run (audio + visual)
Specifications
Capacity 6 x 50 mL
Max. RCF 5,071 x g
Max. Speed 6,000 rpm
Noise Level ≤50 dB (A) with rotor 1137
Electrical Requirements 230 V 50/60 Hz
Product Line EBA 280 S
Height (English) Exterior 9.5 in.
Height (Metric) Exterior 242 mm
Width (English) Exterior 12.8 in.
Width (Metric) Exterior 326 mm
Weight (English) 26.5 lb.
Weight (Metric) 12 kg
Dimensions (HWD) 242 x 326 x 400 mm
Run Time 1 to 99 min., 59 sec., Continuous Run, Short Cycle Mode (Impulse Button)
Frequency 50/60 Hz
Voltage 200/240 V
Depth (Metric) 400 mm
Product Type Centrifuge