Learn More
STEMCELL Technologies EasySep™ Mouse PE Positive Selection Kit II
Immunomagnetic positive selection of mouse cells labeled with PE-conjugated antibodies
1078.50€ - 4866.00€
Especificaciones
Tipo de célula | B Cells, Dendritic Cells, Granulocytes and Subsets, Hematopoietic Stem and Progenitor Cells, Macrophages, Marrow Stromal Cells, Mesenchymal Stem and Progenitor Cells, Monocytes, Mononuclear Cells, Myeloid Cells, NK Cells, Other, Plasma, T Cells |
---|---|
Para utilizar con (aplicación) | Cell Isolation |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
17197741
|
STEMCELL Technologies
17696 |
5 x 109 cells |
4866.00€
1 set |
Fecha estimada de envío: 15-05-2024
Inicie sesión para ver el stock |
|||||
17157731
|
STEMCELL Technologies
17666 |
1 x 109 cells |
1078.50€
1 set |
Fecha estimada de envío: 26-04-2024
Inicie sesión para ver el stock |
|||||
Descripción
- The EasySep™ Mouse PE Positive Selection Kit II is designed to isolate cells that are labeled with PE-conjugated antibodies
- Desired cells are targeted with antibody complexes recognizing PE and dextran-coated magnetic particles
- Labeled cells are separated using an EasySep™ magnet without the use of columns
- Cells of interest remain in the tube while unwanted cells are poured off
- This kit replaces the EasySep™ Mouse PE Positive Selection Kit (Catalog #18554)
Especificaciones
B Cells, Dendritic Cells, Granulocytes and Subsets, Hematopoietic Stem and Progenitor Cells, Macrophages, Marrow Stromal Cells, Mesenchymal Stem and Progenitor Cells, Monocytes, Mononuclear Cells, Myeloid Cells, NK Cells, Other, Plasma, T Cells | |
Cell Isolation |