Learn More
Invitrogen™ EasySelect™ Pichia Expression Kit
Product Code. 10664803
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
Product Code. Quantity Unit Size
10664803 1 Kit Each
1 options
This item is not returnable. View return policy
Product Code. 10664803 Supplier Invitrogen™ Supplier No. K174001
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Fecha estimada de envoi: 01-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
The EasySelect™ Pichia Expression Kit provides all needed components for protein production in the yeast Pichia pastoris.
The EasySelect™ Pichia Expression Kit provides all needed components for protein production in the yeast Pichia pastoris. The EasySelect™ Kit contains Pichia yeast strains, expression vectors, and reagents that enable transformation of Pichia cells. Selection with Zeocin™ antibiotic makes it easy to screen for high-copy number transformed variants.
Advantages of the Pichia pastoris expression system:
  • Culture Pichia cells as easily as E. coli
  • Obtain higher cell density than other microbial expression systems.
  • Generate more protein per cell than other microbial expression systems.
  • Benefit from eukaryotic protein processing, protein folding, and post-translational modifications.
  • Easily scale-up from shake flasks up to industrial-scale fermentation.
A Proven Protein Expression System
Over the past 30 years, Pichia pastoris has been used by labs and industry around the world for producing hundreds of different proteins from many species including human (Ref 1, 2, 3). Pichia pastoris may be grown as easily as bacteria (E. coli) or yeast (Saccharomyces cerevisiae) yet offers many advantages over those systems (Ref 4). Pichia pastoris can grow to much higher cell densities than bacteria or other yeast. Pichia pastoris is capable of growing on methanol as a sole carbon source making scale-up to industrial levels relatively inexpensive.
The Power of the AOX1 Promoter
The EasySelect™ Pichia Expression Kit uses the promoter from the Pichia alcohol oxidase 1 (AOX1) gene to drive production of your protein. Pichia pastoris is capable of growing on methanol as its only carbon source. When grown on methanol as sole carbon source, AOX1 mRNA represents up to 5% of total mRNA and AOX1 enzyme can be up to 30% of cell weight (Ref 5).
The EasySelect™ Pichia Expression system is supplied with the pPICZ and pPICZα vectors. The pPICZ and pPICZα vectors contain a multiple cloning site (MCS) and allow the insertion of your gene of interest under control of the AOX1 promoter. The pPICZ and pPICZα vectors also contain a c-Myc tag for easy detection and a polyhistidine (6xHis) tag for easy detection and purification of your protein.
Since Pichia pastoris naturally secretes few proteins, you can use the extracellular signaling sequence in the pPICZα vectors to force secretion of your protein into the culture medium to simplify down-stream purification.
Get High-Copy Number Integration with Zeocin™ Antibiotic
The EasySelect™ Pichia Expression Kit utilizes the Zeocin™ antibiotic to select for transformed Pichia cells. pPICZ and pPICZα vectors contain the bleomycin gene from Streptoalloteichus hindustanus (Sh ble) that confers resistance to Zeocin™ antibiotic. The pPICZ and pPICZα vectors can be stably integrated into the Pichia genome. By increasing the concentration of Zeocin™ antibiotic, you can easily select for clones with the highest number of integrants and highest protein expression levels.
The EasySelect™ Pichia Expression Kit contains the following components:
  • Stab vials of Pichia pastoris and E. coli strains for starting your cultures.
  • Solutions for making transformation-competent Pichia cells.
  • pPICZ and pPICZα vectors for fusing your gene to the AOX1 promoter.
  • Sequencing primers for confirming insertion of your gene in the pPICZ vectors.
  • Zeocin™ antibiotic for selecting transformed Pichia cells.
References
  1. Cereghino JL, Cregg JM. Heterologous protein expression in the methylotrophic yeast Pichia pastoris. FEMS Microbiol Rev. 2000 Jan;24(1):45-66. [PubMed]
  2. Cereghino GP, Cereghino JL, Ilgen C, Cregg JM. Production of recombinant proteins in fermenter cultures of the yeast Pichia pastoris. Curr Opin Biotechnol. 2002 Aug;13(4):329-32. [PubMed]
  3. Cregg JM. Introduction: distinctions between Pichia pastoris and other expression systems. Methods Mol Biol. 2007;389:1-10. [PubMed]
  4. Cregg JM, Tolstorukov I, Kusari A, Sunga J, Madden K, Chappell T. Expression in the yeast Pichia pastoris. Methods Enzymol. 2009;463:169-89. [PubMed]
  5. Cregg JM, Madden KR, Barringer KJ, Thill GP, Stillman CA. Functional characterization of the two alcohol oxidase genes from the yeast Pichia pastoris. Mol Cell Biol. 1989 Mar;9(3):1316-23. [PubMed]
TRUSTED_SUSTAINABILITY
Specifications
Antibiotic Resistance Bacterial Zeocin™ (ZeoR)
Bacterial or Yeast Strain GS115, KM71H, X-33
Content And Storage The EasySelect Pichia Expression Kit includes 20 μg each of the plasmids pPICZ A, B, and C and pPICZα A, B, and C; Pichia pastoris strains X-33 (wild-type), GS115 (his4), KM71H (arg4 aox1::ARG4), and control strains for both intracellular and secreted expression; 250 mg Zeocin™ antibiotic; media (YP and YNB), EasyComp™ Pichia Transformation Kit, 2 μg each 5' AOX1, 3' AOX1, and α-factor sequencing primers. The pPICZ and pPICZα vectors are also available separately. A lacZ positive expression control is provided with pPICZ.
Store strains at +4°C. Store vectors and primers at -20°C. All components are guaranteed stable for 6 months when properly stored., Store strains at +4°C. Store vectors and primers at -20°C. All components are guaranteed stable for 6 months when properly stored.
Cloning Method Restriction Enzyme⁄ MCS
Promoter AOX1
Cell Type Yeast Cells
Format Kit
Species P. pastoris
Vector pPIC
Expression Mechanism Cell-Based Expression
Expression System Yeast
For Use With (Application) Protein Expression
Product Line EasySelect
Product Type Expression Kit
Protein Tag His Tag (6X), c-Myc Epitope Tag, His Tag (6x), c-Myc Epitope Tag
Quantity 1 Kit
Selection Agent (Eukaryotic) Zeocin
Show More Show Less
compliance-icons
Product Identifier
  • EASYSELECT PICHIA EXPRESSION KIT 00
Signal Word
  • Warning
Hazard Category
  • Germ cell mutagenicity Category 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H341-Suspected of causing genetic defects.
  • H373-May cause damage to organs.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P314-Get medical advice/attention if you feel unwell.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Ethylene glycol, Phleomycin from Streptomyces verticillus
For Research Use Only. Not for use in diagnostic procedures.