missing translation for 'onlineSavingsMsg'
Learn More

E2F8 Antibody, Novus Biologicals™

Goat Polyclonal Antibody

Marca:  Bio-Techne NBP2-89176

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18215427

  • 512.95€ / 0.10 miligramo
Fecha estimada de envío: 21-08-2024
para ver el stock

Descripción

Descripción

E2F8 Polyclonal specifically detects E2F8 in Human, Mouse samples. It is validated for Western Blot, Peptide ELISA.
Especificaciones

Especificaciones

E2F8
Polyclonal
Unconjugated
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA
E2F family member 8, E2F transcription factor 8, E2F-8, FLJ23311, transcription factor E2F8
Peptide with sequence C-KSIESDRRKINSAPS corresponding to internal region according to NP_078956.2.
0.1 mg
Cell Cycle and Replication
79733
Store at -20°C. Avoid freeze-thaw cycles.
IgG
Western Blot, ELISA
0.5 mg/ml
Western Blot 1:100 - 1:2000, Peptide ELISA Detection limit 1:32000
NP_078956.2
Goat
Immunogen affinity purified
RUO
Primary
Human, Mouse
Purified
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales