missing translation for 'onlineSavingsMsg'
Learn More
Learn More
E-Cadherin / CD324 Mouse, Clone: SPM381, enQuireBio™
Mouse Monoclonal Antibody
Marca: EnquireBio 999-MSM5X-P1
Detalles adicionales : Peso : 0.01000kg
Especificaciones
E-Cadherin / CD324 | |
Cell Surface | |
SPM381 | |
Unconjugated | |
NP_001304114 | |
Mouse | |
120-80kDa (Mature) ;135kDa (Precursor) | |
Protein A/G Purified | |
RUO | |
999 | |
IgG1 κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
CDH1 | |
Recombinant human full-length E-Cadherin protein | |
LS174T, Raji, HT29, SK-BR3 cells. Prostate or Colon carcinomas. | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |