missing translation for 'onlineSavingsMsg'
Learn More

Dystrophin Antibody (DMD/3241), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody

Marca:  Novus Biologicals NBP2-79910V

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18216337

  • 578.82€ / 0.10 ml
Fecha estimada de envío: 14-08-2024
para ver el stock

Descripción

Descripción

Dystrophin Monoclonal specifically detects Dystrophin in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Especificaciones

Especificaciones

Dystrophin
Monoclonal
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
DMD
A recombinant fragment (around aa 114-263) of human DMD protein (exact sequence is proprietary) (Uniprot: P11532)
0.1 mL
Cell Biology, Cellular Markers
Human
IgG1 κ
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
DMD/3241
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
BMDDXS272, CMD3B, DXS142, DXS164, DXS164, DXS206, DXS230, DXS239, DXS268, DXS269, DXS270, DXS272, DXS206, DXS230, DXS239, DXS268, DXS269, DXS270, dystrophin, dystrophin (muscular dystrophy, Duchenne and Becker types), includes DXS142
Mouse
Protein A or G purified
RUO
Primary
Store at 4°C in the dark.
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales