Learn More
Invitrogen™ DYRK2 Recombinant Human Protein
DYRK2 is also known as dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 2. Recombinant full-length human DYRK2 was expressed in E. coli cells using an N-terminal GST tag.
Marca:  Invitrogen™ PV6333
Detalles adicionales : Peso : 0.01000kg
 Ver más versiones de este producto
Código de producto. 15448973
  • 5530.00€ / 1 miligramo
Fecha de envío estimada
del proveedor 05-07-2019
Añadir a la cesta
Descripción
Descripción
DYRK2 belongs to a family of protein kinases whose members are presumed to be involved in cellular growth and/or development and is defined by structural similarity of their kinase domains and their capability to autophosphorylate on tyrosine residues. DYRK2 expressed in E. coli demonstrated tyrosine autophosphorylation and catalyzed phosphorylation of histones H3 and H2B in vitro. DYRK2 also regulates p53 to induce apoptosis in response to DNA damage.
  • Validated in LanthaScreen™ Kinase Binding Assay
  • Protocols available for simple use and optimized performance
  • Multiple pack sizes available for convenient use
Especificaciones
Especificaciones
Target & Lead Identification & Validation|Kinase Biology|Kinase Proteins & Expression Reagents|Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis|Industrial & Applied Science|Pharma & Biopharma
92.9kDa
Store at -68 to -85°C. Aliquot at first use to avoid multiple freeze-thaws.
Kinase Assay
Yes
50mM Tris-HCl pH 7.5, 150mM NaCl, 0.25mM DTT, 10mM glutathione, 0.1mM EDTA, 0.1mM PMSF, 25% glycerol
1mg
FLJ21217, FLJ21365
Human
GST Tag
Videos