Learn More
Thermo Scientific™ DyLight™ Free Acids Fluorescent Dyes
Use these non-activated, carboxylic acid forms of DyLight Dyes as controls and calibration standards in fluorescence imaging applications.
Marca:  Thermo Scientific™ 62284
Detalles adicionales : Peso : 0.99000kg
 Ver más versiones de este producto
Código de producto. 10450270
  • 226.00€ / 1 miligramo
Fecha de envío estimada
del proveedor 09-05-2019
Añadir a la cesta
Descripción
Descripción
Thermo Scientific™ DyLight Fluor Free Acids are non-activated forms of selected DyLight Fluorescent Dyes for use as controls and calibration factors in fluorecence imaging applications. The non-activated dyes contain the native carboxyl group of the standard dye molecule.
General DyLight Fluorophore Highlights:
  • Bright fluorescence — intense emission provides superior sensitivity and requires less conjugate
  • Narrow emission spectra — negligible bleed-through between fluorophore channels enables multi-color detection
  • Excellent photostability — exceptional resistance to photobleaching enables fluorescence imaging under the most demanding conditions (e.g., structured illumination, 4Pi microscopy)
  • Buffer stability — conjugated fluorophores are completely stable at pH 4-9
  • Instrument compatibility — excitation and emission spectra correspond with filter sets and laser settings of all popular fluorescence instrumentation
Especificación
Especificación
DyLight™ 800, Free Acid, 1mg, Solid, DyLight™ 800 Dye, 777/794nm Excitation/Emission Wavelength, <4°C Storage Temperature, Used for Protein Biology, Protein Labeling and Crosslinking, Protein Labeling
Powder
DyLight™ 800
DyLight™ 800, Free Acid
1mg
794nm
Unmodified (carboxylate) proprietary near-infrared fluorescent dye
Protein labeling and crosslinking, protein biology
1 to several uses depending on the application
<4°C
777nm
Videos
Documentación