missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DUSP9 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 10826-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
The protein encoded by this gene is a member of the dual specificity protein phosphatase subfamily. These phosphatases inactivate their target kinases by dephosphorylating both the phosphoserine/threonine and phosphotyrosine residues. They negatively regulate members of the mitogen-activated protein (MAP) kinase superfamily (MAPK/ERK, SAPK/JNK, p38), which is associated with cellular proliferation and differentiation. Different members of the family of dual specificity phosphatases show distinct substrate specificities for various MAP kinases, different tissue distribution and subcellular localization, and different modes of inducibility of their expression by extracellular stimuli. This gene product shows selectivity for members of the ERK family of MAP kinases and is localized to the cytoplasm and nucleus. Aberrant expression of this gene is associated with type 2 diabetes and cancer progression in several cell types. Alternate splicing results in multiple transcript variants.Especificaciones
DUSP9 | |
Polyclonal | |
Unconjugated | |
DUSP9 | |
dual specificity phosphatase 9, DUSP9, MAP kinase phosphatase 4, MKP 4, MKP4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1852, 75590 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q99956 | |
DUSP9 | |
DUSP9 Fusion Protein Ag1259 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |