Learn More
DWK Life Sciences DURAN™ Watch Glass Dish, fused rim
Product Code. 11786592 Shop All DWK Life Sciences Products
Change view
Click to view available options
Diameter (Metric):
40 mm
50 mm
60 mm
80 mm
100 mm
125 mm
150 mm
200 mm
250 mm
Unit Size:
Each
Pack of 10
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Unit Size
11786592 40 mm Pack of 10
11796592 50 mm Pack of 10
11706602 60 mm Pack of 10
11716602 80 mm Pack of 10
11726602 100 mm Pack of 10
11756612 125 mm Pack of 10
11766612 150 mm Pack of 10
11716153 200 mm Pack of 10
10202121 250 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Product Code. 11786592 Supplier DWK Life Sciences Supplier No. 213212408
  • ONLINE EXCLUSIVE PRICE
    111.00€
    102.40€ / paquete de 10
    Ahorro 8.60€
    7% Descuento
Fecha estimada de envoi: 04-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
The concave shape of the watch glass dish allow working in very small quantities (e.g. for pretreatments) and covering laboratory glassware with wide openings.
The multitude of different work steps in the laboratory environment demands a multitude of universally applicable glassware. These glass devices usually have a very simple shape and are characterized by their versatility. Watch glasses, for example, are concave curved. The small trough created in this way allows liquids or solids to be filled in.
  • Reduced risk of injury due to fused rim
Specifications
Autoclavable Autoclavable
Material Borosilicate Glass (3.3)
For Use With (Application) Weighing; Covering; Pretreatments; Drying of materials; Available in Boro 3.3 or soda-lime glass
No. per Pack 10/Pk.
Diameter (Metric) 40 mm