Learn More
Lenz Laborglasinstrumente™ Duran Test Tube Sprayers
Product Code. 10772952
Change view
Click to view available options
Capacity (Metric):
12 mL
Unit Size:
Each
Product Code. Capacity (Metric) Unit Size
10772952 12 mL Each
1 options
This item is not returnable. View return policy
Product Code. 10772952 Supplier Lenz Laborglasinstrumente™ Supplier No. 05471200
  • ONLINE EXCLUSIVE PRICE
    72.50€
    58.18€ / paquete de 1
    Ahorro 14.32€
    19% Descuento
Fecha estimada de envoi: 28-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
Capacity (Metric) 12 mL
Material Duran Glass
Quantity 1/Pk.
Type Test Tube Sprayer
Includes Test tube and sprayer head, with clamp and Rubber bulb