Learn More
Hirschmann™ DURAN™ Class AS Micro Burettes Blue Graduations
Product Code. 13138009
Change view
Click to view available options
Capacity (Metric):
2 mL
5 mL
10 mL
Stopcock:
Lateral
Straight
Stopcock Material:
Glass
PTFE
Unit Size:
Pack of 2
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Stopcock Stopcock Material Unit Size
13138009 2 mL Lateral Glass Pack of 2
13168009 2 mL Lateral PTFE Pack of 2
13148009 5 mL Lateral Glass Pack of 2
13178009 5 mL Lateral PTFE Pack of 2
13118009 5 mL Straight Glass Pack of 2
13158009 10 mL Lateral Glass Pack of 2
13188009 10 mL Lateral PTFE Pack of 2
13128009 10 mL Straight Glass Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Product Code. 13138009 Supplier Hirschmann™ Supplier No. 3310140
  • ONLINE EXCLUSIVE PRICE
    447.00€
    358.00€ / paquete de 2
    Ahorro 89.00€
    19% Descuento
Fecha estimada de envoi: 14-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Excellent for dispensing measured amounts of chemical solutions in the laboratory. Manufactured from durable DURAN™ glass tubing.
  • Schellbach
  • Bang pattern
  • Main point ring graduation
  • With blue graduation
  • Acc. to DIN EN ISO 385
  • Support base
  • Conformity certified, with dated batch identification
Specifications
Capacity (Metric) 2 mL
Material Borosilicate Glass
Graduations 0.01 mL
Includes Bang, Main Point Ring Graduation, Schellbach Stripes, Socket
Plug Material Glass
Scale Blue
Stopcock Lateral
Stopcock Material Glass
Tolerance ±0.01 mL
Type Microburet
Class Class AS
Certifications/Compliance DIN EN ISO 385