missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ DSU Recombinant Protein

Human DSU full-length ORF recombinant protein with GST-tag at N-terminal

335.00€ - 508.00€

Especificaciones

Número de acceso AAH32747
Para utilizar con (aplicación) Antibody Production, Protein Array, ELISA, Western Blot
Formulación 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
ID de gen (Entrez) 55686
Peso molecular 49.28
Ver más especificaciones

Products 2
Product Code Brand Cantidad Price Quantity & Availability  
Product Code Brand Cantidad Price Quantity & Availability  
16181683
missing translation for 'viewDocuments' Promotion Details
Abnova™
H00055686-P01.10UG
10 ug
335.00€
10 microgramos
View alternative products
missing translation for 'toSaveUpto' 
»
Fecha estimada de envío: 30-05-2024
para ver el stock
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
16191683
missing translation for 'viewDocuments' Promotion Details
Abnova™
H00055686-P01.25UG
25 ug
508.00€
25 microgramos
View alternative products
missing translation for 'toSaveUpto' 
»
Fecha estimada de envío: 30-05-2024
para ver el stock
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
Description

Description

  • Theoretical MW (kDa): 49.28
  • Preparation method: In vitro wheat germ expression system
  • Purification: Glutathione Sepharose 4 fast flow
  • Storage buffer: 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer

Sequence: MGLRDWLRTVCCCCGCECLEERALPEKEPLVSDNNPYSSFGATLVRDDEKNLWSMPHDVSHTEADDDRTLYNLIVIRNQQAKDSEEWQKLNYDIHTLRQVRREVRNRWKCILEDLGFQKEADSLLSVTKLSTISDSKNTRKAREMLLKLAEETNIFPTSWELSERYLFVVDRLIALDAAEEFFKLARRTYPKKPGVPCLADGQKELHYLPFPSP

Best use within three months from the date of receipt of this protein.

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Specifications

AAH32747
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
49.28
8
Glutathione Sepharose 4 Fast Flow
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MREG
Human
Recombinant
Solution
Antibody Production, Protein Array, ELISA, Western Blot
55686
DSU (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
MGLRDWLRTVCCCCGCECLEERALPEKEPLVSDNNPYSSFGATLVRDDEKNLWSMPHDVSHTEADDDRTLYNLIVIRNQQAKDSEEWQKLNYDIHTLRQVRREVRNRWKCILEDLGFQKEADSLLSVTKLSTISDSKNTRKAREMLLKLAEETNIFPTSWELSERYLFVVDRLIALDAAEEFFKLARRTYPKKPGVPCLADGQKELHYLPFPSP
DSU/FLJ10116/MGC90296/WDT2
MREG
Wheat Germ (in vitro)
GST
Videos
SDS
Documents

Documents

Certificates