Learn More
Drierite™, With indicator, 8 mesh, Honeywell™ Fluka™
Catalog Number 15642400
Click to view available options
Cantidad:
454 g
2.3 kg
12 x 454 g
Envase:
Fiber Drum
Glass Bottle
7778-18-9
CaO4S
136.13
MFCD00010912
OSGAYBCDTDRGGQ-UHFFFAOYSA-L
24497
CHEBI:31346
calcium sulfate
[Ca++].[O-]S([O-])(=O)=O
7778-18-9
CaO4S
136.13
MFCD00010912
OSGAYBCDTDRGGQ-UHFFFAOYSA-L
24497
CHEBI:31346
calcium sulfate
[Ca++].[O-]S([O-])(=O)=O
With indicator, 8 mesh, Drierite™ is a trademark of W. A. Hammond Drierite Company, Ltd.
  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Nombre del producto químico o material Calcium Sulfate
Nombre de nota With indicator
Material Calcium Sulfate
Cantidad 12 x 454 g
Fórmula lineal CaSO4
Tamaño de malla 8mesh
Número UN UN3077
Envase Fiber Drum
Formula Weight (peso de la fórmula) 136.14g/mol