missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ DRG1 (Human) Recombinant Protein

Human DRG1 full-length ORF ( AAH19285, 1 a.a. - 367 a.a.) recombinant protein with GST-tag at N-terminal.

Marca:  Abnova™ H00004733-P01.10ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16198261

  • 302.00€ / 10 microgramos


Fecha estimada de envío
19-10-2020
Añadir a la cesta

Descripción

Descripción

  • Sequence: MSSTLAKIAEIEAEMARTQKNKATAHHLGLLKARLAKLRRELITPKGGGGGGPGEGFDVAKTGDARIGFVGFPSVGKSTLLSNLAGVYSEVAAYEFTTLTTVPGVIRYKGAKIQLLDLPGIIEGAKDGKGRGRQVIAVARTCNLILIVLDVLKPLGHKKIIENELEGFGIRLNSKPPNIGVKKKDKGGINLTATCPQSELDAETVKSILAEYKIHNADVTLRSDATADDLIDVVEGNRVYIPCIYVLNKIDQISIEELDIIYKVPHCVPISAHHRWNFDDLLEKIWDYLKLVRIYTKPKGQLPDYTSPVVLPYSRTTVEDFCMKIHKNLIKEFKYALVWGLSVKHNPQKVGKDHTLEDEDVIQIVKK
Especificaciones

Especificaciones

AAH19285
developmentally regulated GTP binding protein 1
125% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DRG1
GST
4733
Wheat germ expression system
10 μg
DKFZp434N1827, NEDD3
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
Videos
Documentos