missing translation for 'onlineSavingsMsg'
Learn More

Drebrin 1 Antibody (DBN1/2880), Janelia Fluor™ 646, Novus Biologicals™

Mouse Monoclonal Antibody

Marca:  Novus Biologicals NBP2-79902JF646

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18215108

  • 578.82€ / 0.10 ml
Fecha estimada de envío: 19-08-2024
para ver el stock

Descripción

Descripción

Drebrin 1 Monoclonal specifically detects Drebrin 1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Especificaciones

Especificaciones

Drebrin 1
Monoclonal
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
DBN1
Recombinant fragment (aa 150-281) of human Drebrin 1 protein (exact sequence is proprietary) (Uniprot: Q16643)
0.1 mL
Neuroscience
Human
IgG1 κ
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
DBN1/2880
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
D0S117E, Developmentally-regulated brain protein, DKFZp434D064, drebrin, drebrin 1, drebrin E, drebrin E2, drebrin-1
Mouse
Protein G purified
RUO
Primary
Store at 4°C in the dark.
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales