Learn More
Dräger™ Tubos de detección a corto plazo: alcohol
Detector tubes
Marca:  Dräger™ 8101631
Detalles adicionales : Peso : 0.07380kg
 Ver más versiones de este producto
Código de producto. 10029220
Este producto se ha descatalogado y ya no está en venta. Consulte el producto para saber si se ofrecen alternativas o póngase en contacto con nuestro equipo de Atención al Cliente en el número 902 239 303.
Descripción
Descripción
Glass detector tubes are filled with an inert reagent carrier material (silica gel, aluminum oxide, or aluminum silicate) impregnated with an indicating reagent. Reagent produces a colorimetric indication in the presence of a particular gas, vapor, or aerosol. Concentrations can be read directly from the color change on the tube's printed scale. Tubes are hermetically sealed. Most have a two-year shelf life (when stored at room temperature). Available in packs of ten.
Note: Some tests require a pretube or comparison tube in addition to the detector tube. These tubes are included in the box of 10, thus reducing the number of detector tubes. The actual number of detector tubes is cited in the ordering charts.
Order Info
Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.
Especificaciones
Especificaciones
Tubos del detector
15 mg/l
5 min
10
Alcohol 25/a
Alcohol
De 25 a 5000 (metanol), de 50 a 4000 (i-propanol)
10
De 15 ° a 30 °C
Videos