Learn More
Dräger™ CMS Chips: Nitrous Gases
Product Code. 10158041
Change view
Click to view available options
Measurement Range:
0.5 to 15
Response Time:
40 to 350 sec.
Humidity Range:
1 to 40mg/L
Unit Size:
Each
Product Code. Measurement Range Response Time Humidity Range Unit Size
10158041 0.5 to 15 40 to 350 sec. 1 to 40mg/L Each
1 options
This item is not returnable. View return policy
Product Code. 10158041 Supplier Dräger™ Supplier No. 6406060
  • ONLINE EXCLUSIVE PRICE
    570.84€
    195.91€ / paquete de 1
    Ahorro 374.93€
    65% Descuento
Fecha estimada de envoi: 04-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Suitable for 10 measurements of one gas
Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.
Specifications
Air Pressure 700 to 1100 hpa
Type Nitrous Gases CMS Chip
Humidity Range 1 to 40mg/L
Measurement Range 0.5 to 15
Response Time 40 to 350 sec.
Temperature Range (English) 32°F to 104°F
Temperature Range (Metric) 0°C to 40°C