Learn More
Dräger™ CMS Chips: Propane
Product Code. 12374639
Change view
Click to view available options
Measurement Range:
100 to 2000
Unit Size:
Each
Product Code. Measurement Range Unit Size
12374639 100 to 2000 Each
1 options
This item is not returnable. View return policy
Product Code. 12374639 Supplier Dräger™ Supplier No. 6406310
  • ONLINE EXCLUSIVE PRICE
    244.95€
    202.75€ / paquete de 1
    Ahorro 42.20€
    17% Descuento
Fecha estimada de envoi: 11-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Suitable for 10 measurements of one gas
Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.
Specifications
Air Pressure 700 to 1100 hpa
Type Propane CMS Chip
Humidity Range 1 to 40mg/L
Measurement Range 100 to 2000
Response Time 60 to 360 sec.
Temperature Range (English) 32°F to 104°F
Temperature Range (Metric) 0°C to 40°C