Learn More
Dräger™ CMS Chips: Phosgene
Product Code. 12334639
Change view
Click to view available options
Measurement Range:
0.05 to 2
Unit Size:
Each
Product Code. Measurement Range Unit Size
12334639 0.05 to 2 Each
1 options
This item is not returnable. View return policy
Product Code. 12334639 Supplier Dräger™ Supplier No. 6406340
  • ONLINE EXCLUSIVE PRICE
    296.07€
    245.54€ / paquete de 1
    Ahorro 50.53€
    17% Descuento
Fecha estimada de envoi: 04-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Suitable for 10 measurements of one gas
Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.
Specifications
Air Pressure 700 to 1100 hpa
Type Phosgene CMS Chip
Humidity Range 1 to 30mg/L
Measurement Range 0.05 to 2
Response Time 90 to 420 sec.
Temperature Range (English) 32°F to 104°F
Temperature Range (Metric) 0°C to 40°C