Learn More
Dräger™ CMS Chips: Methylene Chloride
Product Code. 12334629
Change view
Click to view available options
Measurement Range:
20 to 400
Unit Size:
Each
Product Code. Measurement Range Unit Size
12334629 20 to 400 Each
1 options
This item is not returnable. View return policy
Product Code. 12334629 Supplier Dräger™ Supplier No. 6406510
  • ONLINE EXCLUSIVE PRICE
    269.45€
    222.62€ / paquete de 1
    Ahorro 46.83€
    17% Descuento
Fecha estimada de envoi: 18-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Suitable for 10 measurements of one gas
Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.
Specifications
Air Pressure 700 to 1100 hpa
Type Methylene Chloride CMS Chip
Humidity Range 1 to 30mg/L
Measurement Range 20 to 400
Response Time 180 to 600 sec.
Temperature Range (English) 50°F to 104°F
Temperature Range (Metric) 10° to 40°C